Learn More
Gibco™ RPMI 1640 Medium

RPMI 1640 Medium was originally developed to culture human leukemic cells in suspension and as a monolayer.
£9.90 - £37.25
Specifications
Cell Line | HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas |
---|---|
Cell Type | Leukemic Cells |
Classification | Animal Origin-free |
Concentration | 1 X |
Form | Liquid |
Requires: Supplementation
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15460564
![]() |
Gibco™
11875101 |
100 mL |
£9.90
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15364138
![]() |
Gibco™
11875-093 |
500 mL |
£20.33
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15450564
![]() |
Gibco™
11875085 |
1000 mL |
£37.25
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPMI 1640 Medium was originally developed to culture human leukemic cells in suspension and as a monolayer. Roswell Park Memorial Institute (RPMI) 1640 Medium has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of RPMI 1640 Medium modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This RPMI is modified as follows:
With: L-glutamine, Phenol Red
Without: HEPES
The complete formulation is available.
Using RPMI
RPMI 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 Medium contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 Medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 Medium uses a sodium bicarbonate buffer system (2.0 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas | |
Animal Origin-free | |
Liquid | |
Standard Serum Supplementation | |
Sterile-filtered | |
No HEPES, No Sodium Pyruvate | |
Gibco™ |
Leukemic Cells | |
1 X | |
RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) | |
Sterile-filtered | |
Glutamine, Phenol Red | |
cGMP-compliant under the ISO 13485 standard | |
Room Temperature |
Safety and Handling
missing translation for 'shelfLife' : 12 Months From Date of Manufacture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.