missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ RPMI 1640 Medium, GlutaMAX™ Supplement, HEPES

Originally developed to culture human leukemic cells in suspension and as a monolayer

£219.00 - £401.00

Specifications

Form Liquid
Without Additives No Sodium Pyruvate
View More Specs

Requires: Supplementation


Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11310902
View Documents
Gibco™
72400153
10 L
£401.00
Each
Please to purchase this item. Need a web account? Register with us today!
11300902
View Documents
Gibco™
72400146
5 L
£219.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RPMI 1640 Medium (Roswell Park Memorial Institute 1640 medium) has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas.

  • Modified as follows: with GlutaMAX™, phenol red, HEPES
  • Contains reducing agent glutathione and high concentrations of vitamins
  • Contains biotin, vitamin B12 and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium
  • Vitamins inositol and choline are present in very high concentrations
  • GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format
  • Contains no proteins, lipids, or growth factors
  • Requires supplementation, commonly with 10% Fetal Bovine Serum (FBS)
  • Uses sodium bicarbonate buffer system (2.0g/L), and therefore requires a 5-10% CO2 environment to maintain physiological pH

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Liquid
No Sodium Pyruvate
Videos
SDS
Documents

Documents

Product Certifications

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.