missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Schneider's Drosophila Medium
GREENER_CHOICE

Modified to contain L-glutamine and sodium bicarbonate

Brand:  Gibco™ 21720024

100.30 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11590576

  • £118.00 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Schneider's Drosophila Medium was originally designed for the growth of S2 cells from the fruit fly, Drosophila melanogaser. Schneider's Drosophila Medium can also be used to culture other dipteran cell lines. This medium can be used for transient or stable transfection and large scale protein production in S2 cells.

Formulation

  • Contains unique components required for S2 cell culture, including malic acid, fumaric acid, alpha-ketoglutaric acid, succinic acid, and the sugar trehelos
  • Commonly supplemented with 10% fetal bovine serum (FBS) before use
  • Requires ambient CO2 in order to maintain physiological pH

Cell Culture, Drosophila S2 Cell Culture, Insect Cell Culture

Compliance

Gibco Schneider's Drosophila Medium is manufactured at a cGMP-compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Insect Cell
Schneider's Drosophila Medium
No HEPES, No Phenol Red, No Sodium Pyruvate
500 mL
S2 (Schneider 2)
Liquid
Glutamine
Gibco™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.