missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Sf-900™ II SFM
GREENER_CHOICE

Product Code. p-4924984
Click to view available options
Quantity:
500 mL
1000 mL
6 x 1 L
Unit Size:
1 L
1 set
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11560366

Brand: Gibco™ 10902096

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells

Sf-900 II SFM is a serum-free, protein-free insect cell culture medium for large-scale production of recombinant proteins expressed using the baculovirus expression vector system (BEVS). This medium is suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines.

  • Superior long-term, high-density growth
  • Optimized for recombinant protein production
  • Serum-free, protein-free, ready-to-use formulation
  • Scalable in bioreactors

Bioproduction, Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture, Viral Vaccine Production

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Sf21, Sf9
Cell Type Insect Cell
Classification Protein-free, Serum-free
Form Liquid
Product Type Insect Cell Serum Free Medium (SFM)
Serum Level Serum-free
With Additives Glutamine
Product Line Gibco, Sf-900
Quantity 500 mL
Shipping Condition Room Temperature
Species S. frugiperda, Spodoptera frugiperda
Content And Storage Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.