missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Sf9 cells in Sf-900™ III SFM

Uused to isolate and propagate recombinant baculoviral stocks and to produce recombinant proteins

Brand:  Gibco™ 12659017

Product Code. 10210523

  • £520.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Alert:

Review the Material Safety Data Sheet before handling.

This item is not returnable. View return policy

Description

Description

Gibco Sf9 cells are adapted to serum-free suspension culture in Gibco Sf-900 III SFM.

  • Recombinant protein expression from a variety of expression systems
  • Good growth in adherent or suspension culture
  • Small, regular size that generates even monolayers and plaques
  • Documented lineage from a low passage Master Cell Bank
  • Quality and performance testing

Baculovirus Production and Titering, Bioproduction, Cell Culture, Insect Cell Culture, Insect Expression, Insect Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Sf9 and Sf21 Cell Culture, Viral Vaccine Production

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 x 1.5 mL (1 x 107 cells/mL)
Storage conditions: liquid nitrogen (vapor phase)
Shipping conditions: frozen
Insect Cells
Sf-900 III SFM (Serum-Free Media)
Insect Cells
Dry Ice
Sf9
S. frugiperda
Sf-900™
1 Vial
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.