Learn More
Gibco™ Distilled Water

High-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents
Brand: Gibco™ 15230162
21.49 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
This distilled water is high-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents. Available in many sizes to meet your needs, this distilled water features:
- Cell culture quality
- Proven performance
- Suitable for use in cell culture and molecular biology applications
Cell culture quality
This water is prepared by distillation and filtration (0.1 μm), with no added substances. We monitor the water quality on a routine basis. Conductivity, total organic carbon, endotoxin, and bioburden are tested daily; metals are tested monthly.
Proven performance
The performance and consistency expected from Gibco™ liquid media products depends on the high-quality water we use in manufacturing. This water is also bottled as Gibco™ Distilled Water.
Use in cell culture and molecular biology applications
This distilled water can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers.
Dual-site cGMP manufacturing and quality system
This water is produced at a cGMP-compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical product made in our Grand Island facility (15230-162). This facility is also registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Order Info
Shipping Condition: Room Temperature

Specifications
Cell Culture | |
Sterile-filtered | |
500 mL | |
Room Temperature |
Bottle | |
Not DEPC-Treated | |
Membrane-Filtered |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2562523 | Certificate of Analysis | 31/01/2025 | 15230279,15230147,15230170,15230261,15230162,15230253 |
2756265 | Certificate of Analysis | 29/01/2025 | 15230279,15230147,15230170,15230261,15230162,15230253 |
2661106 | Certificate of Analysis | 29/01/2025 | 15230279,15230147,15230170,15230261,15230162,15230253 |
2510007 | Certificate of Analysis | 29/01/2025 | 15230279,15230147,15230170,15230261,15230162,15230253 |
2517864 | Certificate of Analysis | 29/01/2025 | 15230279,15230147,15230170,15230261,15230162,15230253 |
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
0Hours
3Minutes
25Seconds
12 -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
30Hours
3Minutes
25Seconds
12
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.