missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Distilled Water
GREENER_CHOICE

Product Code. 15697708
Click to view available options
Quantity:
500 mL
1000 mL
10 x 500 mL
6 x 1 L
Unit Size:
1000mL
500mL
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 15697708

Brand: Gibco™ 15230147

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents

This distilled water is high-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents. Available in many sizes to meet your needs, this distilled water features:

  • Cell culture quality
  • Proven performance
  • Suitable for use in cell culture and molecular biology applications

Cell culture quality

This water is prepared by distillation and filtration (0.1 μm), with no added substances. We monitor the water quality on a routine basis. Conductivity, total organic carbon, endotoxin, and bioburden are tested daily; metals are tested monthly.

Proven performance

The performance and consistency expected from Gibco™ liquid media products depends on the high-quality water we use in manufacturing. This water is also bottled as Gibco™ Distilled Water.

Use in cell culture and molecular biology applications

This distilled water can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers.

Dual-site cGMP manufacturing and quality system

This water is produced at a cGMP-compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical product made in our Grand Island facility (15230-162). This facility is also registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

pH 7
Grade Cell Culture
Packaging Bottle
Sterility Sterile-filtered
Treatment(s) Not DEPC-Treated
Quantity 1000 mL
Purification Method Membrane-Filtered
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.