missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ TrypLE™ Express Enzyme (1X), no phenol red
GREENER_CHOICE

Product Code. 11528856
Click to view available options
Quantity:
100 mL
500 mL
5 L
20 x 100mL
Unit Size:
100mL
5 L
500mL
Pack of 20
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11528856

Brand: Gibco™ 12604021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Unlike porcine trypsin, free of animal-derived components

Cleaves peptide bonds on the C-terminal sides of lysine and arginine, and is a direct replacement for trypsin. Exceptional purity increases specificity and reduces damage to cells that can be caused by other enzymes present in some trypsin extracts. Used for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells.

  • Gentle on cells
  • Room temperature-stable for 24 months
  • Easy to use
  • Animal origin-free
  • Gentle on cells, purity increases specificity due to the action of a single enzyme and reduces damage caused by cleavage from multiple enzymes in trypsin and other extracts
  • Can be directly substituted for trypsin in existing protocols
  • Dilution alone inactivates TrypLE Express, avoiding the need for trypsin inhibitors, such as FBS

Energy efficient, less hazardous, sustainable packaging

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured at a cGMP-compliant facility that is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Chelators EDTA
Content And Storage Storage conditions: 15°C to 30°C, protect from light
Alternate storage conditions: 2°C to 8°C or -5°C to -20°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture
Cell Type Mammalian Cells
Classification Animal Origin-free
Concentration 1 X
Culture Type Adherent Cell Culture
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile-filtered
Without Additives No Phenol Red
For Use With (Application) Clinical Research, Translational Research
Green Features Energy efficient, Sustainable packaging
Osmolality Check and Record
Product Line Gibco
Quantity 500 mL
Remarks Room Temperature Stable
Shipping Condition Room Temperature
Source Animal origin-free
Tested For In vitro Bioassay
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.