missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Trypsin-EDTA (0.05%), phenol red
GREENER_CHOICE

Product Code. 11580626
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11580626

Brand: Gibco™ 25300054

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas.

Due to its digestive strength, trypsin is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation. The trypsin concentration required for dissociation varies with cell type and experimental requirements.

  • Documented traceability
  • Dual-site cGMP Manufacturing
  • Quality testing
  • Solutions tested for pH, osmolality, sterility, and performance
  • Raw materials are verified for e-beam irradiation and tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal, and viral contamination, as well as multiple activity assays, ash analysis, and moisture analysis
  • Documented traceability available including lot traceability, animal origin certificates, lot analyses, irradiation certificates, viral inactivation summary, and supply chain transparency
TRUSTED_SUSTAINABILITY

Specifications

Chelators EDTA
Content And Storage Storage conditions: -5°C to -20°C
Shelf life: 24 months from date of manufacture
Cell Type Mammalian Cells
Classification Animal Origin
Concentration 1 X
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Phenol Red, EDTA
Osmolality 270 - 310 mOsm/kg
pH 7.1 to 8
Product Line Gibco
Quantity 100 mL
Shipping Condition Wet ice
Source Animal Origin
Tested For In vitro Bioassay
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.