Learn More
Gibco™ Trypsin-EDTA (0.05%), phenol red
Made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas
Brand: Gibco™ 25300062
48.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 0.50000kg
Description
Due to its digestive strength, trypsin is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation. The trypsin concentration required for dissociation varies with cell type and experimental requirements.
- Documented traceability
- Dual-site cGMP Manufacturing
- Quality testing
- Solutions tested for pH, osmolality, sterility, and performance
- Raw materials are verified for e-beam irradiation and tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal, and viral contamination, as well as multiple activity assays, ash analysis, and moisture analysis
- Documented traceability available including lot traceability, animal origin certificates, lot analyses, irradiation certificates, viral inactivation summary, and supply chain transparency
Specifications
EDTA | |
Mammalian Cells | |
1 X | |
Cell Culture Dissociation Reagent | |
Phenol Red, EDTA | |
270 - 310 mOsm/kg | |
Gibco™ | |
Wet Ice | |
In vitro Bioassay |
Storage conditions: -5°C to -20°C Shelf life: 24 months from date of manufacture |
|
Animal Origin | |
Liquid | |
Sterile-filtered | |
Sustainable packaging | |
7.1 to 8 | |
500 mL | |
Animal Origin |