Learn More
Gibco™ Trypsin-EDTA (0.25%), phenol red

Beschreibung
Due to its digestive strength, trypsin is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation. The trypsin concentration required for dissociation varies with cell type and experimental requirements.
- Solutions tested for pH, osmolality, sterility, and performance
- Raw materials are verified for e-beam irradiation and tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal, and viral contamination, as well as multiple activity assays, ash analysis, and moisture analysis
- Documented traceability available including lot traceability, animal origin certificates, lot analyses, irradiation certificates, viral inactivation summary, and supply chain transparency
Cell Culture, Mammalian Cell Culture
Bestellinformation
Shipping Condition: Dry Ice
Compliance
Manufactured at a cGMP compliant facility registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards.
Spezifikation
Spezifikation
| Chelators | EDTA |
| Content And Storage | Storage conditions: -5°C to -20°C Shipping conditions: Wet ice Shelf life: 24 months from date of manufacture |
| Cell Type | Mammalian Cells |
| Classification | Animal Origin |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | Cell Culture Dissociation Reagent |
| Sterility | Sterile-filtered |
| Sterilization Method | Sterile-filtered |
| With Additives | Phenol Red, EDTA |
| Mehr anzeigen |
Sicherheit und Handhabung
- Trypsin, 0.25% (1X) with EDTA 4Na, liquid
- Danger
- Respiratory sensitiser Category 1
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : trypsin
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.