missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Zeocin™ Selection Reagent
Selection antibiotic, formulation of phleomycin D1, a glycopeptide antibiotic produced by Streptomyces verticillus
£361.00 - £1560.00
Specifications
Concentration | 100 mg/mL |
---|---|
Culture Type | Mammalian Cell Culture, Insect Cell Culture |
Form | Liquid |
Product Type | Antibiotic |
Sterility | Sterile-filtered |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11508976
|
Gibco™
R25005 |
50 mL |
£1560.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10072492
|
Gibco™
R25001 |
8 x 1.25 mL |
£361.00
Pack of 8 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Gibco™ Zeocin™ Selection Antibiotic is a formulation of phleomycin D1, a glycopeptide antibiotic produced by Streptomyces verticillus
- Zeocin™ Selection Antibiotic acts by binding to and cleaving DNA, leading to cell death
- It is used for selection of of mammalian, insect, yeast, bacterial, and plant cell cultures
- Working concentration ranges between 50–400 μg/mL depending on the cell type
- This product is supplied at 100 mg/mL in sterile water, in 8 x 1.25 mL vials
Order Info
Shipping conditions: Ice
Specifications
100 mg/mL | |
Liquid | |
Sterile-filtered | |
Zeocin™ |
Mammalian Cell Culture, Insect Cell Culture | |
Antibiotic | |
Eukaryotic Selection/Stable Cell Line Generation, Bacterial Selection |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title