missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Giemsa Stain Product Code.: 11414667

Thermo Scientific Chemicals Giemsa Stain

Product Code. 11414667
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11414667

Brand: Thermo Scientific Chemicals B21172.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Giemsa Stain is used as a laboratory reagent and stain for microscopy. It is also used for the histopathologic detection of malaria and other parasites such as spirochete and protozoan. The stain is also been incorporated in differential staining between bacterial and human cells. The compound has been used in combination with May-Grunwald or Wright Stain (sc-203763) as a histological hematology stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Giemsa Stain is used as a laboratory reagent and stain for microscopy. It is also used for the histopathologic detection of malaria and other parasites such as spirochete and protozoan. The stain is also been incorporated in differential staining between bacterial and human cells. The compound has been used in combination with May-Grunwald or Wright Stain (sc-203763) as a histological hematology stain.

Solubility
Insoluble in water (<0.1%), methanol (5.8 mg/L), and ethanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51811-82-6
Molecular Formula C14H14ClN3S
Molecular Weight (g/mol) 291.80
MDL Number MFCD00012112,MFCD00081642
InChI Key NALREUIWICQLPS-UHFFFAOYSA-N
Synonym azure a, dimethylthionine, giemsa stain, azur a, n,n-dimethylthionine, 3-amino-7-dimethylamino phenothiazin-5-ium chloride, methylene azure a, giemsa solution, 3-amino-7-dimethylaminophenazathionium chloride, unii-m731v243efShow MoreShow Less
PubChem CID 13735
IUPAC Name (7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride
SMILES [Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1

Specifications

Melting Point ∼300°C
Quantity 50 g
Solubility Information Insoluble in water (<0.1%),methanol (5.8mg/L),and ethanol.
Chemical Name or Material Giemsa Stain

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252689 Certificate TypeCertificate of Analysis Date15/11/2024 Catalog NumberB21172.09,B21172.18
Lot Number10252144 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog NumberB21172.09,B21172.18
Lot Number10245681 Certificate TypeCertificate of Analysis Date10/02/2023 Catalog NumberB21172.09,B21172.18
Lot Number10245387 Certificate TypeCertificate of Analysis Date17/01/2023 Catalog NumberB21172.09,B21172.18
Lot Number10234421 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21172.09,B21172.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Giemsa Stain >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.