Learn More
Epredia™ Giemsa Stain
Take advantage of an excellent classical blood film stain used on peripheral blood smears and bone marrow with the Epredia™ Giemsa Stain.
£224.00
Specifications
Type Giemsa Stain
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12583687
View Documents Promotion Details
Epredia™
89004
1 gal.
£224.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 22-07-2024
to see stock.
Description
Description
  • Erythrocytes stain pink with platelets exhibiting a very light, pale hue
  • Leukocyte nuclear chromatin stains magenta-purple
  • Monocyte cytoplasm stains the classical “ground glass” blue
  • Lymphocyte cytoplasm stains clear, sky blue
Specifications
Specifications
Giemsa Stain