missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilac™ Gilactiv™ Bowls with Handles, 50 Cycle
Gilactiv™ 50 Cycle Bowls with Handles are reusable plastic bowls treated with the exclusive Sanitized™ technology, which protects the bowl's surface against viruses and bacteria.
Brand: Gilac™ G627490
Additional Details : Weight : 0.65000kg
Description
- Gilactiv™ reusable plastic bowl treated with the exclusive Sanitized™ technology, which protects the bowl's surface against viruses and bacteria.
- Proven antibacterial and antiviral effectiveness on the bowl's surface (empty or full) for a broad spectrum of germs, including HCoV-229E human coronavirus.
- This article contains a biocidal product with the active substance: Silver Phosphate Glass.
- Tests conducted by an EU-approved French independent laboratory showed that 98.6% of the virus was wiped out after a two-hour period.
- Choose Gilactiv™ enhanced protection.
- Test results available on request.
- Outstanding durability even in the intensive use cycle.
- Ideal for institutional catering use.
- Pack of 10.
Specifications
Bowl | |
Blue | |
65 mm | |
0.65 kg |
HDPE | |
134 mm | |
10 Pk. |