missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilac™ Gilactiv™ Container with Lid
Gilactiv™ Container with Lid is treated with the exclusive Sanitized™ technology, which protects the container's surface against viruses and bacteria.
Brand: Gilac™ G119990
Additional Details : Weight : 3.05000kg
Description
- Proven antibacterial and antiviral effectiveness on the container's surface (empty or full) for a broad spectrum of germs, including HCoV-229E human coronavirus.
- This article contains a biocidal product with the active substance: Silver Phosphate Glass.
- Tests conducted by an EU-approved French independent laboratory showed that 98.6% of the virus was wiped out after a two-hour period.
- Choose Gilactiv™ enhanced protection.
- Test results available on request.
- No gaps nor overlaps and no sharp corners.
- Rewritable identification cartridge for inventory tracking.
- Recessed handles for better handling.
- Smooth interior walls and rounded corners for easier cleaning.
- Machine washable.
- Compatible with all GILAC 4 and 6-wheel dollies.
Specifications
Storage Tote | |
Blue | |
14.5 gal. |
HDPE | |
Rectangular | |
55 L |