Learn More
Gilac™ Gilactiv™ Tumblers, 22 Cycle, Blue
Gilactiv™ 22 Cycle Tumblers are reusable plastic tumblers treated with the exclusive Sanitized™ technology, which protects the tumbler's surface against viruses and bacteria.
Brand: Gilac™ G627290
Description
- Gilactiv™ reusable plastic tumbler treated with the exclusive Sanitized™ technology, which protects the tumbler's surface against viruses and bacteria.
- Proven antibacterial and antiviral effectiveness on the tumbler's surface (empty or full) for a broad spectrum of germs, including HCoV-229E human coronavirus.
- This article contains a biocidal product with the active substance: Silver Phosphate Glass.
- Tests conducted by an EU-approved French independent laboratory showed that 98.6% of the virus was wiped out after a two-hour period.
- Choose Gilactiv™ enhanced protection.
- Test results available on request.
- Outstanding durability even in the intensive use cycle.
Specifications
Plastic | |
7.4 oz. | |
Tumbler Cup |
0.22 L | |
10 Pk. |