missing translation for 'onlineSavingsMsg'
Learn More

Oakton™ Glass Beads for Mills

Product Code. 15237649 Shop All Cole-Parmer Products
Click to view available options
Diameter (Metric):
0.1 to 0.15 mm
0.5 mm
1 mm
Unit Size:
1kg
Each
This item is not returnable. View return policy

Product Code. 15237649

Brand: Oakton™ 3627050

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Suitable glass beads for the mini-BeedBeater™ Mill. Cole-Parmer™ Glass Beads for Mills are used to crush and separate small samples from bone to microbial spores.

  • Available in three different sizes
  • Crush samples such as bone and microbial spores
  • Handles small samples efficiently—from 100μL to 1μL
  • Non-foaming, non-aerosol method preserves enzymes, nucleic acids, and organelles

Specifications

Type Beads
Material Glass
Diameter (Metric) 1 mm
For Use With (Equipment) Mini-Bead Beater Glass Mill
Quantity 1 lb.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.