Learn More
Gilson™ PIPETMAN™ Pipettes
GREENER_CHOICE
PIPETMAN brings a new level of comfort into Gilson range with a redesigned piston mechanism that reduces the force needed to aspirate and dispense liquids. PIPETMAN maintains the trusted durability, accuracy, and reliability of the original PIPETMAN pipettes with comfort.
Brand:  Gilson™ F144055M
250.52 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
 View more versions of this product
Product Code. 12366122
  • £274.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Body (handle) in PVDF minimizes the hand-warming effect during pipetting and is resistant against aggressive chemicals
  • Direct volume reading in decimal digits helps prevent mistakes in volume setting
  • Comes with classic metal or plastic tip ejector
  • Clip ejector fits securely into the body of the pipette and helps avoid slipping over time
  • Redesigned seal, lighter ejection springs and high quality piston grease offer as low as 2 Newtons of force to pipet
  • High quality piston grease
Model Specific Features:
  • Models P2 and P10 incorporate dual position tip ejectors for use with D10 (short) and DL10 (long) tips
Order Info
Trade in program available. Please contact your Fisher Scientific sales representative for more details.
Specifications
Specifications
±0.025, ±0.075, ±0.1μL
No
Red
Reusable
Ergonomic
P10
Pipetman
Non-sterile
1 to 10 μL
D10, DL10, DF10ST, DFL10ST tip
14.37 in.
12.01 in.
4.72 in.
Red
D10, DL10, DF10ST, DFL10ST Tip
Autoclavable
Manual
D-10, DL10, DF10ST, DFL10ST Tip
PVDF, Stainless Steel
1
No
Pipette
Extremely low pipetting forces, lightweight design
Stainless Steel Tip Ejector
36.5 cm
30.5 cm
12 cm
Certified Quality
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products