Learn More
Gilson™ PIPETMAN™ L Micro-volume Kit
PIPETMAN™ L Micro-volume Kit is designed for molecular biology applications.
Brand: Gilson™ F167550
737.25 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.00000kg
Description
The PIPETMAN L Micro-volume Kit offers the PIPETMAN P2L and P10L, ideal for pipetting enzymes and reagents; and a PIPETMAN P100L, suited for small-volume dispensing of aqueous fluids of moderate viscosity and density (molecular biology, immunology, etc.).
Kit Contents:
- 3 PIPETMAN L Pipettes: P2L, P10L, and P100L with Gilson certificates
- 2 racks of certified-quality PIPETMAN DIAMOND Tips: DL10 and D200
- 3 SINGLE™ pipette holders
- 3 Plastic tip ejectors
- 1 Gilson guide to pipetting
- 1 Two-minute inspection poster
Specifications
Autoclavable | |
Reusable | |
Ergonomic | |
1 | |
Non-sterile | |
0.2 to 100 μL | |
1 Year | |
37 cm | |
31 cm | |
12 cm |
Blue/White | |
Manual Air Displacement Pipette | |
Polypropylene | |
Pipetman | |
Micro-Volume Kit | |
Manual Air Displacement Pipette | |
14.57 in. | |
12.2 in. | |
4.72 in. | |
Certified Quality |