Learn More
Gilson™ PIPETMAN™ M Pipettes
Product Code. 15560490 Shop All Gilson Products
Click to view available options
Volume (Metric):
0.5 to 10 μL
1 to 10 mL
1 to 20 μL
10 to 10 μL
10 to 100 μL
10 to 300 μL
100 to 1200 μL
20 to 200 μL
20 to 300 μL
50 to 1200 μL
500 to 5000 μL
Number of Channels:
1
8
12
Unit Size:
Each
Product Code. 15560490
Brand: Gilson™ F81015
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Single or multichannel. Equipped with motorized piston that virtually eliminates pipetting variability among users and minimizes RSI (Repetitive Strain Injuries) among users
  • Motorized piston with adjustable piston speed that is compatible with varying solution densities
  • Lockable volume to prevent inaccuracies
  • Color-coded push-button corresponds to matching tip packs
  • Service alert reminders
  • Unique tip holder design allows for a secure fit of most brands of tips
  • Features four essential pipetting modes dash (en) reverse, mix, repetitive and pipetting to best fit your application
  • Pipette can be used while charging eliminating bottlenecks and downtime
  • Designed to exert equal pressure on each piston to ensure consistent volume levels across all channels
  • Power carrousel conveniently holds and charges up to 5 PIPETMAN M pipettes at once, charging 80% of its full capacity in less than one hour
  • Gilson Trident and unique spacer distribute ejection forces evenly over each tip, reducing the force required to eject tips
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips D1000, DF1000, D1200, DF1200
Color Blue
Model P1200M
Number of Channels 1
Product Line Pipetman
Type Pipette
Volume (Metric) 100 to 1200 μL
Finger Style Ergonomic
Warranty 1 Year