Learn More
Gilson™ PIPETMAN™ Neo Pipets
Fits most brands of tips. Gilson PIPETMAN™ Neo Pipets are the next generation of the PIPETMAN Classic™ with lower spring and ejection forces.
Brand: Gilson™ F144563
Code : 50
Additional Details : Weight : 0.18000kg
Description
- Redesigned tip ejector and ejection springs reduces tip ejection forces
- Unique clip ejector fits securely into the body of the pipette to avoid slipping over time
- PVDF handle limits the transfer of heat from hand to the pipette and is resistant to aggressive chemicals
- Stainless steel tip ejector resists corrosion and offers a smooth tip ejection stroke
- Color-coded push-button for ease of identification
- Unique tip holder allows for a secure fit of most brands of tip
- Models P2N and P10N incorporate dual position tip ejectors, allowing the use of D10 (short) and DL10 (long)
Specifications
±0.1, ±0.1, ±0.2μL | |
D200, DF30ST | |
1 | |
Pipette | |
Ergonomic |
ISO8655 | |
P20N | |
Pipetman™ Neo™ | |
2 to 20 μL | |
Color coded push button, PVDF handle, Stainless Steel tip ejectors, universal tip holder |