Learn More
Gilson™ PIPETMAN™ TOWERPACK™ Refill System
Differentiates tip models with color-coded racks and indicates the compatible Gilson pipet models. Gilson™ PIPETMAN™ TOWERPACK™ Refill System is available in autoclavable and sterilized versions.
£41.15 - £74.90
Specifications
Filtered | Non-filtered |
---|---|
Color | Clear |
Autoclavable | Autoclavable |
Product Type | Tower Pack™ Pipet Refill System |
Material | Polypropylene |
Product Code | Brand | Volume (Metric) | For Use With (Equipment) | Sterility | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | For Use With (Equipment) | Sterility | Price | Quantity & Availability | |||
10758944
|
Gilson™
F167102 |
0.1 to 20 μL | DL10 (longer tip) 0.1 to 20μL 45mm L Pipetman™ Diamond™ pipet tips | Non-sterile |
£63.25
Pack of 960 |
In Stock
Log in to see stock. |
|||
10021902
|
Gilson™
F167104 |
100 to 1000 μL | D1000 100 to 1000μL 66mm L Pipetman™ Diamond™ pipet tips | Non-sterile |
£47.15
Pack of 672 |
In Stock
Log in to see stock. |
|||
10113532
|
Gilson™
F167101 |
0.1 to 10 μL | D10 0.1 to 10μL 30mm L Pipetman™ Diamond™ pipet tips | Non-sterile |
£69.90
Pack of 960 |
In Stock
Log in to see stock. |
|||
10011902
|
Gilson™
F167103 |
2 to 200 μL | D200 2 to 200μL 51mm L Pipetman™ Diamond™ pipet tips | Non-sterile |
£55.85
Pack of 960 |
In Stock
Log in to see stock. |
|||
10387372
|
Gilson™
F167201 |
0.1 to 10 μL | D10 0.1 to 10μL 30mm L Pipetman™ Diamond™ pipet tips | Sterile |
£74.90
Pack of 960 |
Estimated Shipment: 13-09-2024
Log in to see stock. |
|||
10317522
|
Gilson™
F167105 |
20 to 300 μL | D300 20 to 300μL 51mm L Pipetman™ Diamond™ pipet tips | Non-sterile |
£55.85
Pack of 960 |
In Stock
Log in to see stock. |
|||
10347422
|
Gilson™
F167202 |
0.1 to 20 μL | DL10 0.1 to 20μL 45mm L Pipetman™ Diamond™ pipet tips | Sterile |
£61.20
Pack of 960 |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||
10718564
|
Gilson™
F167203 |
2 to 200 μL | D200 2 to 200μL 51mm L Pipetman™ Diamond™ pipet tips | Sterile |
£67.35
Pack of 960 |
In Stock
Log in to see stock. |
|||
10235452
|
Gilson™
F167204 |
100 to 1000 μL | D1000 200 to 1000μL 66mm L Pipetman™ Diamond™ pipet tips | Sterile |
£41.15
Pack of 672 |
In Stock
Log in to see stock. |
|||
10728564
|
Gilson™
F167205 |
20 to 300 μL | D300 20 to 300μL 51mm L Pipetman™ Diamond™ pipet tips | Sterile |
£59.60
Pack of 960 |
In Stock
Log in to see stock. |
|||
Description
- Color-coded racks to differentiate tip models and indicate the compatible Gilson™ pipet models
- Available in autoclavable and sterilized versions
- Fits Universal Reload box (Cat. No. F167100G, available separately)
- Sterilized Tower Pack is individually wrapped in medical device-quality plastic to help protect tips from contamination
Spécification
Non-filtered | |
Autoclavable | |
Polypropylene |
Clear | |
Tower Pack™ Pipet Refill System |