missing translation for 'onlineSavingsMsg'
Learn More

Ginkgolide B, MedChemExpress

Product Code. p-200101583
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16484887

Brand: Medchem Express HYN0784

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ginkgolide B (BN-52021), an important active terpenoid from Ginkgo biloba leaves, is reported to increase cell viability and decrease cell apoptosis.

Ginkgolide B (BN-52021), an important active terpenoid from Ginkgo biloba leaves, is reported to increase cell viability and decrease cell apoptosis.

Specifications

Chemical Name or Material Ginkgolide B
CAS 15291-77-7
Color White
Purity Grade Notes Research
Molecular Formula C20H24O10
Quantity 50 mg
Synonym BN-52021
Solubility Information DMSO : ≥ 100 mg/mL (235.63 mM)
SMILES O[C@@H]1C(C2([C@H]3O)[C@](OC3=O)([H])O4)([C@@](O5)([H])C[C@H]2C(C)(C)C)C4(C5=O)[C@@](O)([C@@H]6C)[C@@]1([H])OC6=O
Molecular Weight (g/mol) 424.4
Formula Weight 424.4
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.