missing translation for 'onlineSavingsMsg'
Learn More

Girard's Reagent T, 99%

Product Code. p-3776729 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 10326750

Brand: Thermo Scientific 119870500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-46-6 | C5H14ClN3O | 167.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-46-6
Molecular Formula C5H14ClN3O
Molecular Weight (g/mol) 167.64
MDL Number MFCD00012009
InChI Key YSULOORXQBDPCU-UHFFFAOYSA-N
Synonym girard's reagent t, girard reagent t, girard t reagent, girards reagent t, betaine hydrazide hydrochloride, unii-oo3mf0us46, trimethylaminoacetohydrazide chloride, carbazoylmethyltrimetylammonium chloride, trimethylacethydrazide ammonium chloride, trimethylammonium chloride acethydrazide
PubChem CID 67156
SMILES [Cl-].C[N+](C)(C)CC(=O)NN

Specifications

Assay Percent Range 99%
Linear Formula H2NNHCOCH2N(CH3)3Cl
Fieser 01,410
Merck Index 15, 4465
IUPAC Name (2-hydrazinyl-2-oxoethyl)-trimethylazanium;chloride
Formula Weight 167.64
Packaging Glass bottle
Quantity 50 g
Chemical Name or Material Girard's Reagent T
compliance-icons
Product Identifier
  • Girard''s Reagent T
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2-hydrazino-N,N,N-trimethyl-2-oxoethanaminium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.