missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GKN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563479
This item is not returnable.
View return policy
Description
Immunogen sequence: PPLNNLQWYI YEKQALDNMF SSKYTWVKYN PLESLIKDVD WFLLGSPIEK LCKHIPLYKG EVVENT Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 65%.
The function of this protein remains unknown.
Specifications
| GKN2 | |
| Polyclonal | |
| Unconjugated | |
| GKN2 | |
| 1810036H07Rik; Blot; Blottin; BRICD1B; BRICHOS domain containing 1B; Down-regulated in gastric cancer; down-regulated in gastric cancer GDDR; gastrokine 2; gastrokine-2; GDDR; Gkn2; PRO813; RGD1311934; TFIZ1; trefoil factor interactions(z) 1; UNQ465/PRO813; VLTI465 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 200504 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86XP6 | |
| GKN2 | |
| Recombinant protein corresponding to Human GKN2. Recombinant protein control fragment (Product #RP-99699). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction