missing translation for 'onlineSavingsMsg'
Learn More

Corning™ GL45 Polypropylene Screw Caps

Product Code. 17243484
Click to view available options
Color:
Gray
Green
Light Gray
Purple
For Use With (Equipment):
Storage bottle
No. per Case:
10/Cs.
20/Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

Linerless plug seal design for efficient sealing

These are various sizes and types of screw caps for use with storage bottles.

TRUSTED_SUSTAINABILITY

Specifications

Color Purple
Material Polypropylene
No. per Case 20/Cs.
No. per Pack 20/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 45
Closure Color Purple
Autoclavable Autoclavable
For Use With (Equipment) Storage bottle
Closure Material Polypropylene
Closure Type Plug Seal
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'