missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Edmund Bühler GmbH™ Glass Beads
For the dissolution of samples into powder
£43.45 - £85.60
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10041041
|
Edmund Bühler GmbH™
0001 064 |
0.13 mm |
£49.65
1kg |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
10526332
|
Edmund Bühler GmbH™
0001 063 |
0.1 mm |
£60.35
1kg |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
10115751
|
Edmund Bühler GmbH™
0001 066 |
0.50 mm |
£43.45
1kg |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
10648841
|
Edmund Bühler GmbH™
0001 065 |
0.30 mm |
£85.60
1kg |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
Description
- Glass construction
- PbO-free
- 1kg each in plastic bottle
- For dissolution of samples into powder
Specifications
Glass | |
PbO-free |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title