Learn More
Hirschmann™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation are manufactured from durable DURAN™ glass tubing.
Brand:  Hirschmann™ 2820192
 View more versions of this product
Product Code. 10689451
  • £52.00 / Each
Estimated Shipment: 22-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS ground and polyethylene stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Specifications
DIN EN ISO 1042
1000 mL
300 mm
24/29 NS Standard Ground
Yes
Polyethylene
Borosilicate Glass, Duran Glass
±0.6 mL
±0.6 mL
A