Learn More
Hirschmann™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation are manufactured from durable DURAN™ glass tubing.
Brand:  Hirschmann™ 2820160
Additional Details : Weight : 0.04500kg
 View more versions of this product
Product Code. 11708071
  • £31.60 / Pack of 2
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Description
Description
  • With NS ground and polyethylene stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Specifications
DIN EN ISO 1042
10 mL
90 mm
7/16 NS Standard Ground
Yes
Polyethylene
Borosilicate Glass, Duran Glass
±0.025 mL
±0.025 mL
A