Learn More
Hirschmann™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation are manufactured from durable DURAN™ glass tubing.
Brand:  Hirschmann™ 2820152
 View more versions of this product
Product Code. 11782412
  • £31.95 / Pack of 2
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS ground and polyethylene stopper
  • Acc. to DIN ENO ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Specifications
DIN EN ISO 1042
5 mL
70 ± 5 mm
0.025 mL
A
Polyethylene
Borosilicate Glass
7/16 NS Standard Taper Ground
Yes