Learn More
Fisherbrand™ Glass Body pH Electrode, S7
General purpose, standard, glass body pH Meter with an S7 connector. Fisherbrand™ Glass Body pH Electrode, S7 uses a KCl 4M + AGCl electrolyte.
Brand:  Fisherbrand™ 277B504-160-S7
Product Code. 11786338
  • £65.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • General purpose
  • Glass body
  • pH range: 0 - 14
  • Electrolyte: KCl 4M + AgCl
  • Connector: S7
Specifications
Specifications
S7
160 mm
0°C to 80°C
12 mm
0 to 14
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Glass Body pH Electrode, S7 > Connector Type: S7