missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Glass Bottom for Desiccator
Description
- Bottom for desiccator
Certifications
ISO 13 130, DIN 12 491
Specifications
143 mm | |
I.D.: 91 mm | |
Desiccator (Type: 150mm) |
Bottom | |
Glass | |
158 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction