Learn More
Fisherbrand™ Glass Conical Separating Funnel with Glass Stopcock
Manufactured from Borosilicate Glass and complies with ISO 4800 standards. Fisherbraqnd™ Glass Conical Separating Funnel with Glass Stopcock is available with an interchangeable glass or PTFE stopcock, but supplied with a glass and plastic stopper.
Brand: Fisherbrand™
Code : 41
Additional Details : Weight : 0.78020kg
Description
- Manufactured from Borosilicate glass
- Complies with ISO 4800 standards
- Supplied with a glass and plastic stopper
- Available with interchangeable glass or PTFE stopcock
Specifications
1000 mL | |
ISO 4800 | |
Glass | |
29/32 |
Borosilicate Glass | |
Each | |
Glass, Plastic | |
Separating funnel |