Learn More
BRAND™ Glass Discharge Tip
AR-GLAS™ construction. Includes silicone tubing.
Brand:  BRAND™ 11500
Product Code. 11814070
  • £109.00 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Clear AR-GLAS™ construction
  • For automatic burette
  • Schilling pattern
  • Includes silicone tubing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Glass
Discharge Tip
Automatic Burette
Silicone tubing