Learn More
Macherey-Nagel™ Chromabond™ Glass Fiber Frit
Ideal for high temperature SPE applications. Macherey-Nagel™ Chromabond™ Glass Fiber Frit are designed for individual packing of empty glass SPE columns with Chromabond™ adsorbents (glass columns, adsorbents sold separately).
Brand:  Macherey-Nagel™ 730191
 View more versions of this product
Product Code. 10015671
  • £169.00 / Pack of 250
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Frits act as filters in solid phase extraction chromatography. They help to hold the column packing in place and facilitates sorbent packing in the column. Glass fiber columns/frits have many advantages:
  • Resistant to acids, bases and most organic solvents
  • Has low absorption for most biological macromolecules
  • Ideal for high-temperature applications
Specifications
Specifications
Solid Phase Extraction
Frit
Glass Fiber
Glass Column
3 mL
250 Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Chromabond™ Glass Fiber Frit > Volume: 3mL