Learn More
Horst™ Glass Fiber GBW Model Fleecy Tape
Fleecy glass fiber tape for insulating heated sections. Horst™ GBW Glass Fiber Fleecy Tape provides protection up to 450°C.
Brand:  Horst™ 100201
Product Code. 11903925
  • £306.00 / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Easy to apply thermal insulation
  • Tape format
  • Easy to remove if necessary
Specifications
Specifications
3mm thick
GBW
25mm
450°C
30 m
Glass fiber
Fleecy Tape