missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Pneumatic Quick-Disconnect Lockout

Isolates pneumatic energy without the expense and inconvenience of installing in-line lockout valves.

Brand:  Brady™ 065645

Additional Details : Weight : 0.17920kg

Product Code. 15643507

  • £80.90 / Pack of 6
Estimated Shipment: 16-07-2024
to see stock.
Add to basket

Description

Description

  • Device is fitted to the male fitting, isolating equipment from all compressed air sources
  • Accommodates the majority of 6mm, 9mm and 12mm male fittings
Specifications

Specifications

Pneumatic Shutoff
Glass-filled Nylon
6 Pk.
Red
Pneumatic Quick-Disconnect Lockout
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers