missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Glass Funnel Filter with Sintered Glass Disc

Constructed of borosilicate glass and available in a variety of capacities. Fisherbrand™ Glass Funnel Filters with Sintered Glass Disc filter medium precipitates ranging from 16 - 40μm and 10 to 14μm pore sizes respectively.

Brand:  Fisherbrand™

42.48 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.



Code : 40

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 11922148

  • £67.20 / Each
In Stock
to see stock.
Add to basket

Description

Description

Constructed of borosilicate glass and available in a variety of capacities. Fisherbrand™ Glass Funnel Filter with Sintered Glass Disc filters medium precipitates ranging from 16 - 40μm and 10 to 14μm pore sizes respectively.

  • Funnels constructed of borosilicate glass.
  • Funnel filters constructed of sintered glass.
  • Filters medium precipitates with grade 3 and grade 4 porosity.
Specifications

Specifications

80 mL
40mm
3
Borosilicate Glass
Sintered glass disc
Filter Funnel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers