missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Glass Graduated Class AS Type 1 Pipets

Pipet liquids with precision. Fisherbrand™ graduated class AS type 1 pipets are made from soda lime glass. Type 1 pipets have the zero mark at the top.

Brand:  Fisherbrand™

6.44 GBP valid until 2024-07-31
Use promo code "21752" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.05220kg

 View more versions of this product

Product Code. 15287925

  • £9.60 / Pack of 2
In Stock
to see stock.
Add to basket

Description

Description

Pipet liquids with precision. Fisherbrand™ graduated class AS type 1 pipets are made from soda lime glass. Type 1 pipets have the zero mark at the top.

  • Soda lime glass
  • Type 1
  • Blue graduations
  • Class AS
  • Batch Certificate of Compliance is downloadable 
Specifications

Specifications

Soda Lime Glass
0.1 mL
±0.10 mL
Pipet
Yes
5 Pack
2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers