Learn More
BUCHI Glass Hood
Use this hood with the Wet Digestor B-440. BUCHI Glass Hood as part of the Wet Digestor B-440 is designed for parallel pre-incineration of samples.
Brand: BUCHI 036081
Code : A0
Additional Details : Weight : 0.79000kg
Description
- Glass hood for the Wet Digester B-440
- Hot fumes rise into the glass hood and are safely withdrawn by the suction flow of the Scrubber
Specifications
Glass Hood | |
B-440 Wet Digester |