missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Soda Lime Glass Pasteur Pipette
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
150 mm
230 mm
270 mm
Type:
Cotton-plugged Pasteur Pipet
Unplugged Pasteur Pipet
Tip Style:
Closed Tip
Standard Tip
Unit Size:
Pack of 1000
Description
- Pasteur pipettes made of soda-lime glass are a good alternative to plastic pipettes
- Complies with ISO 7712
- Pasteur pipettes with short or long extended, fine tip (150 mm, 230 mm and 270 mm)
- Pasteur pipettes with and without cotton plug
- The filling volume is approx. 2 mL
- Ideal for use with Fisherbrand Pipettors
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | Soda Lime Glass |
| For Use With (Application) | Single Use in Microbiological and Serological Laboratory Applications, Pipetting Liquids, Dropwise Dosing of Liquids and Reagents, Distribution of Heated Liquid Quantities |
| Quantity | 1000 Pk. |
| Tip Style | Standard Tip |
| Volume (English) | 0.07 oz. |
| Volume (Metric) | 2 mL |
| Length (Metric) | 230 mm |
| Type | Cotton-plugged Pasteur Pipet |
| Disposable | Single-use |
| Show More |
Expires: 2025-12-31
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
Expires: 2025-12-31
Waste Water Testing and Analysis Management
Save on essential products for waste water testing and analysis management. Discover top deals and optimize your testing process. Promo code: 24145
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Soda Lime Glass Pasteur Pipette >
Spot an opportunity for improvement?Share a Content Correction