missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Soda Lime Glass Pasteur Pipette

Product Code. 11785098 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
150 mm
230 mm
270 mm
Type:
Cotton-plugged Pasteur Pipet
Unplugged Pasteur Pipet
Tip Style:
Closed Tip
Standard Tip
Unit Size:
Pack of 1000

Product Code. 11785098

Only null left
Add to basket
Add to basket

Soda Lime Glass Pasteur Pipette is intended for single use in microbiological and serological laboratories.

  • Pasteur pipettes made of soda-lime glass are a good alternative to plastic pipettes
  • Complies with ISO 7712
  • Pasteur pipettes with short or long extended, fine tip (150 mm, 230 mm and 270 mm)
  • Pasteur pipettes with and without cotton plug
  • The filling volume is approx. 2 mL
  • Ideal for use with Fisherbrand Pipettors
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Description With constriction and cotton plug
Material Soda Lime Glass
For Use With (Application) Single Use in Microbiological and Serological Laboratory Applications, Pipetting Liquids, Dropwise Dosing of Liquids and Reagents, Distribution of Heated Liquid Quantities
Includes Cotton
Quantity 1000 Pk.
Tip Style Closed Tip
Volume (English) 0.07 oz.
Volume (Metric) 2 mL
Length (Metric) 230 mm
Type Cotton-plugged Pasteur Pipet
Disposable Single-use
For Use With (Equipment) Fisherbrand Pipetters
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.