missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Soda Lime Glass Pasteur Pipette
Soda Lime Glass Pasteur Pipette is intended for single use in microbiological and serological laboratories.
Brand: Fisherbrand™ M4270NW250FS4
87.12 GBP valid until 2024-11-01
Use promo code "21717" to get your promotional price.
Description
- Pasteur pipettes made of soda-lime glass are a good alternative to plastic pipettes
- Complies with ISO 7712
- Pasteur pipettes with short or long extended, fine tip (150 mm, 230 mm and 270 mm)
- Pasteur pipettes with and without cotton plug
- The filling volume is approx. 2 mL
- Ideal for use with Fisherbrand Pipettors
Specifications
Not Autoclavable | |
Single Use in Microbiological and Serological Laboratory Applications, Pipetting Liquids, Dropwise Dosing of Liquids and Reagents, Distribution of Heated Liquid Quantities | |
Standard Tip | |
2 mL | |
Cotton-plugged Pasteur Pipet | |
Fisherbrand Pipetters |
Soda Lime Glass | |
1000 Pk. | |
0.07 oz. | |
270 mm | |
Single-use |