Learn More
Cytiva Glass Plate
Glass plate for use with the miniVE Vertical Electrophoresis System.
Brand: Cytiva SE262P-5
Code : A0
Additional Details : Weight : 0.18900kg
Description
- Glass
- 10 x 10.5 cm
- miniVE Vertical Electrophoresis System Accessory
Specifications
Glass | |
10 x 10.5 cm |