missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Glass Plates for Standard Flatbed Units

Supplied in packs of 3

Brand:  Cytiva 80110687

 View more versions of this product

Product Code. 11852180

  • £600.00 / Pack of 2
Estimated Shipment: 14-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 125 x 240 x 0.5 mm
  • Polystyrene spacer
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard Flatbed Unit
Glass
0.01 in.
200 mm
U-Frame Glass Plate
2/Pk.
0.5 mm
260 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers