missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Hecht™ Glass Raschig Rings
For use with distillation and chemical engineering applications.
£38.40 - £186.00
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10159740
|
Karl Hecht™
41400003 |
3 mm |
£186.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10449601
|
Karl Hecht™
41400010 |
10 mm |
£38.40
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Provided in packs of 1kg
- Available in two different sizes
- Clear glass
Specifications
Raschig Rings | |
1kg pack |