missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Glass Screw Neck Specimen Vials

Store specimens securely with squat or tall form glass vials. Fisherbrand™ Glass Screw Neck Specimen Vials are available in a variety of sizes. Fisherbrand™ polypropylene screw caps are sold separately.

Brand:  Fisherbrand™ 15314789

81.57 GBP valid until 2025-12-31
Use promo code "24350" to get your promotional price.


 View more versions of this product

Product Code. 15314789

  • £144.56 / Pack of 264
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Description

Description

  • Manufactured from clear Type 1B neutral glass
  • Squat and tall form options with standard neck sizes
  • 1 dram equates to approximately 3.55mL (or 0.125 fl. oz.)
  • Suitable for media, diagnostic, storage, display, and sample collection applications
  • Fisherbrand™ Black Polypropylene Screw Caps for Glass Specimen Vials sold separately
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Neutral Glass (Type 1B)
Tall form
72 mm
20 mm
14 mL
Screw Cap
18 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Glass Screw Neck Specimen Vials > Tall form; 14mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.