Learn More
Heidolph instruments Glass Set G5 Coated
Dry ice condenser for low-boiling solvents with transparent plastic coating for added safety that is ideal in most scientific, research, and other laboratories.
Brand:  Heidolph instruments 513-00510-00
Product Code. 10117561
  • £2375.00 / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
    • Glass set for Hei-VAP rotary evaporators
    • Dry Ice condenser for low-boiling solvents
    • With transparent plastic coating for added safety
    • All glassware sets feature GL 10 thread; the inlet tube that can be connected with a threaded fitting
    • Glassware set includes one 1.000-ml evaporating flask and one 1.000-ml receiving flask
    • For use with LABOROTA 4000/4001 efficient, LABOROTA 4010/4011 digital, LABOROTA 4002/4003 control, ABOROTA 4002/4003 control vario
Specifications
Specifications
115/230 VAC
Rotary Evaporator
LABOROTA 4000/4001 Efficient, LABOROTA 4010/4011 Digital, LABOROTA 4002/4003 Control, ABOROTA 4002/4003 Control Vario