Learn More
Heidolph instruments Glass Sheath for Sensor Temperature PT1000
Accessories for Heidolph MR Series Hei-Tec Hotplate Stirrers that are ideal in research and scientific laboratory environments.
Brand: Heidolph instruments
Code : A0
Additional Details : Weight : 0.20000kg
Description
- Accessories for Heidolph MR Series Hei-Tec Hotplate Stirrers
- Pt 1000 Temperature Sensor is glass coated for improved chemical resistance
Specifications
6 x 250 mm |